missing translation for 'onlineSavingsMsg'
Learn More

ER-000444793, MedChemExpress

Product Code. 16499736
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499736 50 mg 50mg
16493188 10 mg 10mg
16450709 1 mg 1mg
16415927 5 mg 5mg
16414476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499736 Supplier Medchem Express Supplier No. HY100852
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ER-000444793 is a potent inhibitor of mitochondrial permeability transition pore (mPTP) opening. ER-000444793 inhibits mPTP with an IC50 of 2.8 μM.

ER-000444793 is a potent inhibitor of mitochondrial permeability transition pore (mPTP) opening. ER-000444793 inhibits mPTP with an IC50 of 2.8 μM.

Specifications

Chemical Name or Material ER-000444793
CAS 792957-74-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H18N2O2
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (141.08 mM; Need ultrasonic)
SMILES O=C(NC1=C(CC2=CC=CC=C2)C=CC=C1)C(C3=CC=CC=C3N4)=CC4=O
Molecular Weight (g/mol) 354.4
Formula Weight 354.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.