missing translation for 'onlineSavingsMsg'
Learn More

(±)-Equol, MedChemExpress

Product Code. 16489646
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489646 50 mg 50mg
16472616 25 mg 25mg
16464386 100 mg 100mg
16440916 200 mg 200mg
16403098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489646 Supplier Medchem Express Supplier No. HY100583A
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (±)-Equol is the racemate of equol. (±)-equol exhibits EC50s of 200 and 74 nM for human ERα and ERβ, respectively. Equol is a metabolite of the soy isoflavones, daidzin and daidzein.

(±)-Equol is the racemate of equol. (±)-equol exhibits EC50s of 200 and 74 nM for human ERα and ERβ, respectively. Equol is a metabolite of the soy isoflavones, daidzin and daidzein.

Specifications

Chemical Name or Material (±)-Equol
CAS 94105-90-5
Color White
Purity Grade Notes Research
Molecular Formula C15H14O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (412.76 mM)
SMILES OC1=CC=C2CC(C3=CC=C(O)C=C3)COC2=C1
Molecular Weight (g/mol) 242.27
Formula Weight 242.27
Percent Purity 99.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.