missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Equine IFN-gamma Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1586-SP
This item is not returnable.
View return policy
Description
IFN-gamma Polyclonal specifically detects IFN-gamma in Equine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IFN-gamma | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3458 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.25-1.25 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| P42160 | |
| IFNG | |
| E. coli-derived recombinant equine IFN-gamma Ala25-Gln166 Accession # P42160 | |
| 25 μg | |
| Primary | |
| Detects equine IFN-gamma in ELISAs and Western blots. In sandwich immunoassays, approximately 20% cross-reactivity with recombinant canine IFN-gamma is observed, less than 6% cross-reactivity with recombinant bovine IFN-gamma and recombinant feline IFN-gamma is observed, and less than 0.2% cross-reactivity with recombinant human IFN-gamma , recombinant mouse IFN-gamma , recombinant rat IFN-gamma , recombinant porcine IFN-gamma , recombinant rhesus macaque IFN-gamma , and recombinant cotton rat IFN-gamma is observed. | |
| Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction