missing translation for 'onlineSavingsMsg'
Learn More

Equilin, MedChemExpress

Product Code. 16488656
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488656 100 mg 100mg
16434567 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488656 Supplier Medchem Express Supplier No. HYB1176
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.

Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.

Specifications

Chemical Name or Material Equilin
CAS 474-86-2
Color White
Purity Grade Notes Research
Molecular Formula C18H20O2
Quantity 100 mg
Synonym 7-Dehydroestrone
Solubility Information DMSO : 100 mg/mL (372.65 mM; Need ultrasonic)
SMILES C[C@]1([C@](CC2)([H])C3=CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O
Molecular Weight (g/mol) 268.35
Formula Weight 268.35
Percent Purity 99.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.