missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Equilibration Buffer EQ-EF

Product Code. 18061091
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18061091 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18061091 Supplier Macherey-Nagel™ Supplier No. 740380.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Equilibration Buffer EQ-EF 1000 mL to be used with NucleoBond.

  • Accessories

Specifications

Content And Storage 1000 mL
Description Buffer
For Use With (Application) Vaccine-grade plasmid DNA isolation
No. of Reactions 1000 mL
Product Type Equilibration Buffer
Quantity 1000 mL
compliance-icons
Product Identifier
  • Buffer EQ-EF (1000 mL)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use suitable extinguisher media to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.