missing translation for 'onlineSavingsMsg'
Learn More

Equilenin Sulfate Sodium Salt (stabilized with TRIS, 50% w/w), TRC

Product Code. 30386052
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386052 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386052 Supplier Toronto Research Chemicals Supplier No. TRCE59241025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Equilenin Sodium Sulfate
CAS 16680-48-1
Molecular Formula C18 H17 O5 S . Na
Quantity 25 mg
Synonym Estra-1,3,5,7,9-pentaen-17-one, 3-(sulfooxy)-, sodium salt (9CI), Estra-1,3,5,7,9-pentaen-17-one, 3-hydroxy-, hydrogen sulfate, monosodium salt (8CI), Equilenin sodium sulfate, Equilenin sulfate sodium salt
InChI Formula InChI=1S/C18H18O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h2-5,10,16H,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t16-,18-;/m0./s1
SMILES [Na+].C[C@]12CCc3c(ccc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CCC2=O
IUPAC Name sodium;[(13S,14S)-13-methyl-17-oxo-12,14,15,16-tetrahydro-11H-cyclopenta[a]phenanthren-3-yl] sulfate
Molecular Weight (g/mol) 368.379
Formula Weight 368.069
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.