missing translation for 'onlineSavingsMsg'
Learn More

EPZ020411 hydrochloride, MedChemExpress

Product Code. p-200097993
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498167

Brand: Medchem Express HY12970A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM, it has >10 folds selectivity for PRMT6 over PRMT1 and PRMT8. EPZ020411 hydrochloride can be used for the research of cancer.

EPZ020411 hydrochloride is a selective inhibitor of PRMT6 with an IC50 of 10 nM, it has >10 folds selectivity for PRMT6 over PRMT1 and PRMT8. EPZ020411 hydrochloride can be used for the research of cancer.

Specifications

Chemical Name or Material EPZ020411 hydrochloride
CAS 2070015-25-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H39ClN4O3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (104.37 mM; Need ultrasonic) ∣H2O : 20 mg/mL (41.75 mM; ultrasonic and warming and heat to 60°C)
SMILES CNCCN(C)CC1=CNN=C1C2=CC=C(O[C@H]3C[C@H](OCCC4CCOCC4)C3)C=C2.Cl[H]
Molecular Weight (g/mol) 479.06
Formula Weight 479.06
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.