missing translation for 'onlineSavingsMsg'
Learn More

EPZ015666, MedChemExpress

Product Code. 16495956
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495956 100 mg 100mg
16491507 50 mg 50mg
16468017 5 mg 5mg
16425308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495956 Supplier Medchem Express Supplier No. HY12727
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM.

EPZ015666 (GSK3235025) is an orally available inhibitor of PRMT5 with an IC50 of 22 nM.

Specifications

Chemical Name or Material EPZ015666
CAS 1616391-65-1
Color White
Purity Grade Notes Research
Molecular Formula C20H25N5O3
Quantity 100 mg
Synonym GSK3235025
Solubility Information DMSO : 100 mg/mL (260.80 mM; Need ultrasonic)
SMILES O=C(NC[C@H](O)CN1CCC(C=CC=C2)=C2C1)C3=CC(NC4COC4)=NC=N3
Molecular Weight (g/mol) 383.44
Formula Weight 383.44
Percent Purity 98.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.