missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPZ011989 trifluoroacetate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
EPZ011989 trifluoroacetate is a potent, selective orally bioavailable EZH2 inhibitor with Ki 3000-fold selectivity over other HMTase.
Specifications
Specifications
| Chemical Name or Material | EPZ011989 trifluoroacetate |
| Physical Form | Solid |
| CAS | 1598383-41-5 |
| Color | Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C37H52F3N5O6 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | EPZ-011989 trifluoroacetate |
| Solubility Information | DMSO : 100 mg/mL (138.92 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction