missing translation for 'onlineSavingsMsg'
Learn More

EPZ004777, MedChemExpress

Product Code. 16426378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426378 10 mg 10mg
16418967 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16426378 Supplier Medchem Express Supplier No. HY15227
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress EPZ004777 is a potent, selective DOT1L inhibitor with an IC50 of 0.4 nM.

EPZ004777 is a potent, selective DOT1L inhibitor with an IC50 of 0.4 nM.

Specifications

Chemical Name or Material EPZ004777
CAS 1338466-77-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H41N7O4
Quantity 10 mg
Solubility Information DMSO : 80 mg/mL (148.24 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](CN(C(C)C)CCCNC(NC4=CC=C(C(C)(C)C)C=C4)=O)O3)C=C2)=NC=N1
Molecular Weight (g/mol) 539.67
Formula Weight 539.67
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.