missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EPZ 015666

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
EPZ 015666 is a potent and selective protein arginine methyltransferase 5 (PRMT5) inhibitor (IC50 = 22 nM, Ki = 5 nM). Displays selectivity for PRMT5 over a panel of 20 other protein methyltransferases. Exhibits dose-dependent antitumor effects in multiple mantle cell lymphoma (MCL) xenograft models. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-[(2S)-3-(3,4-Dihydro-2(1H)-isoquinolinyl)-2-hydroxypropyl]-6-(3-oxetanylamino)-4-pyrimidinecarboxamide |
| CAS | 1616391-65-1 |
| Quantity | 25 mg |
| Molecular Formula | C20H25N5O3 |
| Formula Weight | 383.44 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction