missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPX Rabbit anti-Human, Clone: EPX/3908R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07389-100ug
This item is not returnable.
View return policy
Description
EPX Monoclonal specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| EPX | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.5-1 μg/mL, Immunohistochemistry-Paraffin | |
| EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN | |
| Human eosinophils from a patient with hypereosinophilic syndrome | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| EPX/3908R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 8288 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction