missing translation for 'onlineSavingsMsg'
Learn More

EPX Antibody (rEPO104) - Azide and BSA Free, Novus Biologicals™

Product Code. 18331463 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331463 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331463 Supplier Novus Biologicals Supplier No. NBP308457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EPX Monoclonal specifically detects EPX in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPX
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rEPO104
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence : 0.5-1 ug/mL, Immunohistochemistry-Paraffin
Formulation 10mM PBS
Gene Alias EC 1.11.1.7, eosinophil peroxidase, EPOEPER, EPPEC 1.11.1, EPX-PEN
Host Species Mouse
Immunogen Human eosinophils from a patient with hypereosinophilic syndrome
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8288
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.