missing translation for 'onlineSavingsMsg'
Learn More

Epstein-Barr virus p18 Goat anti-Epstein-Barr virus, Polyclonal, Abnova™

Product Code. 16018287
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16018287 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16018287 Supplier Abnova Supplier No. PAB29841.500uL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat Polyclonal Antibody

Goat polyclonal antibody raised against Epstein-Barr virus (EBV) p18.

  • High specificity
  • High reactivity

Specifications

Antigen Epstein-Barr virus p18
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against Epstein-Barr virus (EBV) p18.
Dilution ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Host Species Goat
Immunogen Recombinant Epstein-Barr virus viral-capsid antigen (EBV-VCA) p18.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Epstein-Barr virus (EBV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.