missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epstein Barr Virus Nuclear Antigen 6 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-66598
This item is not returnable.
View return policy
Description
Epstein Barr Virus Nuclear Antigen 6 Polyclonal specifically detects Epstein Barr Virus Nuclear Antigen 6 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| EpsteinBarrVirusNuclearAntigen6 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| Epstein Barr Virus Nuclear Antigen 3C | |
| Sheep were immunized with recombinant EBNA-3C protein (Uniprot: P03204). | |
| 250 μg | |
| Virus | |
| Purified |
| Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Delipidated, heat inactivated, lyophilized stable whole serum with No Preservative | |
| Sheep | |
| Whole Antiserum | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction