missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epstein Barr Virus Nuclear Antigen 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144513
This item is not returnable.
View return policy
Description
Epstein Barr Virus Nuclear Antigen 3 Polyclonal Antibody for Western Blot, ELISA
Plays an essential role for activation and immortalization of human B-cells. Represses transcription of viral promoters TP1 and Cp through interaction with host RBPJ, and inhibits EBNA2-mediated activation of these promoters. Since Cp is the promoter for all EBNA mRNAs, EBNA3A probably contributes to a negative autoregulatory control loop.
Specifications
| Epstein Barr Virus Nuclear Antigen 3 | |
| Polyclonal | |
| Unconjugated | |
| EBNA3 | |
| EBNA3; Epstein-Barr nuclear antigen 3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 3783762 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.936 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P12977 | |
| EBNA3 | |
| Recombinant Epstein-Barr virus EBNA3 protein (1-138aa). | |
| 100 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction