missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epstein Barr Virus Mouse anti-Virus, Clone: CS2, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Epstein Barr Virus |
|---|---|
| Clone | CS2 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
Description
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Epstein Barr Virus | |
| 0.2 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
| IgG1 κ | |
| Protein A or G purified |
| CS2 | |
| Western Blot 1 to 2 μg/mL, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title