missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epstein-Barr Virus Hhv-4 GP42 Humanized Recombinant Human Monoclonal Antibody (SAA2098)

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA554319

Product Code. 30132331

  • £343.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Compatible with Direct ELISA.

Epstein-Barr Virus HHV-4 GP42 facilitates viral entry by binding to HLA class II molecules on host cells, promoting fusion with the cell membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epstein-Barr Virus Hhv-4 GP42 Humanized
Recombinant Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A/G
RUO
Virus
Antibody
VHH-hFc
ELISA
SAA2098
Unconjugated
BZLF2; EBV Hhv-4 GP42
Recombinant EBV/HHV-4 GP42/Glycoprotein 42.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.