missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epstein Barr Virus Ea-D Monoclonal Antibody (H11B)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA17277

Product Code. 11550422

  • £440.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MA1-7277 antibody reacts with Epstein-Barr virus EA-D in viral and infected tissue samples. MA1-7277 has been successfully used in Western blot, Immunofluorescence and Immunohistochemical applications. The MA1-7277 antibody was raised against EBV EA-D protein.

Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin's disease and Burkitt's Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Epstein Barr Virus Ea-D
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG2
Immunohistochemistry, Western Blot
H11B
Unconjugated
EBV; EBV EA; EBV EAD; EBV early
EBV EA-D.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.