missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epstein Barr Virus Antibody (CS1-4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08501AF405
This item is not returnable.
View return policy
Description
Epstein Barr Virus Monoclonal antibody specifically detects Epstein Barr Virus in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Epstein Barr Virus | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
| Recombinant fusion protein containing the sequence of bacterial beta-galactosidase and the carboxyl half of EBV-encoded LMP | |
| 100 μg | |
| Immune System Diseases, Immunology, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CS1-4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction