missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epsin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | Epsin 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Epsin 1 Polyclonal specifically detects Epsin 1 in Human samples. It is validated for Western Blot.Specifications
| Epsin 1 | |
| Polyclonal | |
| Rabbit | |
| Q9Y6I3 | |
| 29924 | |
| Synthetic peptides corresponding to EPN1(epsin 1) The peptide sequence was selected from the C terminal of EPN1. Peptide sequence AATPTPTPPTRKTPESFLGPNAALVDLDSLVSRPGPTPPGAKASNPFLPG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EH domain-binding mitotic phosphoprotein, EPS-15-interacting protein 1, epsin 1, epsin-1 | |
| EPN1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title