missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epsin 1 Antibody (6F7F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£435.00
Specifications
| Antigen | Epsin 1 |
|---|---|
| Clone | 6F7F9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
Epsin 1 Monoclonal specifically detects Epsin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| Epsin 1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| EH domain-binding mitotic phosphoprotein, EPS-15-interacting protein 1, epsin 1, epsin-1 | |
| EPN1 | |
| IgG1 | |
| Protein A or G purified |
| 6F7F9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 29924 | |
| Purified recombinant fragment of human EPN1 (AA: 106-254) expressed in E. Coli. | |
| Primary | |
| 60.3 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title