missing translation for 'onlineSavingsMsg'
Learn More
Learn More
epsilon-Sarcoglycan Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00008910-D01P
This item is not returnable.
View return policy
Description
epsilon-Sarcoglycan Polyclonal antibody specifically detects epsilon-Sarcoglycan in Human samples. It is validated for Western Blot
Specifications
| epsilon-Sarcoglycan | |
| Polyclonal | |
| Western Blot | |
| NP_003910.1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 8910 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| dystonia 11, myoclonic, DYT11, epsilon-sarcoglycan, Epsilon-SG, ESG, Sarcoglycan epsilon, sarcoglycan, epsilon, SGCE | |
| SGCE (NP_003910.1, 1 a.a. - 437 a.a.) full-length human protein. MQLPRWWELGDPCAWTGQGRGTRRMSPATTGTFLLTVYSIFSKVHSDRNVYPSAGVLFVHVLEREYFKGEFPPYPKPGEISNDPITFNTNLMGYPDRPGWLRYIQRTPYSDGVLYGSPTAENVGKPTIIEITAYNRRTFETARHNLIINIMSAEDFPLPYQAEFFIKNMNVEEMLASEVLGDFLGAVKNVWQPERLNAINITSALDRGGRVPLPINDLKEGVYVMVGADVPFSSCLREVENPQNQLRCSQEMEPVITCDKKFRTQFYIDWCKISLVDKTKQVSTYQEVIRGEGILPDGGEYKPPSDSLKSRDYYTDFLITLAVPSAVALVLFLILAYIMCCRREGVEKRNMQTPDIQLVHHSAIQKSTKELRDMSKNREIAWPLSTLPVFHPVTGEIIPPLHTDNYDSTNMPLMQTQQNLPHQTQIPQQQTTGKWYP | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction