missing translation for 'onlineSavingsMsg'
Learn More

Epsilon-momfluorothrin, MedChemExpress

Product Code. p-200096807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494498

Brand: Medchem Express HY111634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epsilon-momfluorothrin is a type I synthetic pyrethroid insecticide, activates constitutive androstane receptor (CAR), and induces hepatocellular tumors in rats.

Epsilon-momfluorothrin is a type I synthetic pyrethroid insecticide, activates constitutive androstane receptor (CAR), and induces hepatocellular tumors in rats.

Specifications

Chemical Name or Material Epsilon-momfluorothrin
CAS 1065124-65-3
Color White
Purity Grade Notes Research
Molecular Formula C19H19F4NO3
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (12.98 mM; Need ultrasonic)
SMILES O=C([C@H]1C(C)(C)[C@@H]1/C=C(C#N)/C)OCC2=C(F)C(F)=C(COC)C(F)=C2F
Molecular Weight (g/mol) 385.35
Formula Weight 385.35
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.