missing translation for 'onlineSavingsMsg'
Learn More

Epsilon-momfluorothrin, MedChemExpress

Product Code. 16494498
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494498 10 mg 10mg
16443116 25 mg 25mg
16417227 5 mg 5mg
16410817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494498 Supplier Medchem Express Supplier No. HY111634
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Epsilon-momfluorothrin is a type I synthetic pyrethroid insecticide, activates constitutive androstane receptor (CAR), and induces hepatocellular tumors in rats.

Epsilon-momfluorothrin is a type I synthetic pyrethroid insecticide, activates constitutive androstane receptor (CAR), and induces hepatocellular tumors in rats.

Specifications

Chemical Name or Material Epsilon-momfluorothrin
CAS 1065124-65-3
Color White
Purity Grade Notes Research
Molecular Formula C19H19F4NO3
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (12.98 mM; Need ultrasonic)
SMILES O=C([C@H]1C(C)(C)[C@@H]1/C=C(C#N)/C)OCC2=C(F)C(F)=C(COC)C(F)=C2F
Molecular Weight (g/mol) 385.35
Formula Weight 385.35
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.