missing translation for 'onlineSavingsMsg'
Learn More

Epsilon Dipeptide, TRC

Product Code. 30386293
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386293 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386293 Supplier Toronto Research Chemicals Supplier No. TRCE592296250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epsilon Dipeptide
CAS 51513-74-7
Molecular Formula C31 H53 N5 O11
Quantity 250 mg
Synonym L-Lysine, N6-[N2,N6-bis[(1,1-dimethylethoxy)carbonyl]-L-lysyl]-N2-[(1,1-dimethylethoxy)carbonyl]-, 2,5-pyrrolidinedione derivative
InChI Formula InChI=1S/C31H53N5O11/c1-29(2,3)44-26(41)33-19-13-10-14-20(34-27(42)45-30(4,5)6)24(39)32-18-12-11-15-21(35-28(43)46-31(7,8)9)25(40)47-36-22(37)16-17-23(36)38/h20-21H,10-19H2,1-9H3,(H,32,39)(H,33,41)(H,34,42)(H,35,43)/t20-,21-/m0/s1
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-6-[[(2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoyl]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
Molecular Weight (g/mol) 671.779
Formula Weight 671.374
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.