missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactone, 99%

Product Code. 11474043
Change view
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474043 2500 g 2500g
11488950 100 g 100g
11464043 500 g 500g
11484043 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11474043 Supplier Thermo Scientific Alfa Aesar Supplier No. A10299.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 502-44-3 | C6H10O2 | 114.144 g/mol

ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

Solubility
Miscible with water and most organic solvents.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Combustible liquid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-44-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -2°C
Density 1.078
Boiling Point 235°C to 236°C
Flash Point 109°C (228°F)
Assay Percent Range 99%
Refractive Index 1.463
Quantity 2500 g
Beilstein 106919
Solubility Information Miscible with water and most organic solvents.
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.