missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS8L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20461-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.Specifications
| EPS8L2 | |
| Polyclonal | |
| Unconjugated | |
| EPS8L2 | |
| EPS8 like 2, EPS8 like protein 2, EPS8 related protein 2, EPS8L2, EPS8R2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64787, 98845 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H6S3 | |
| EPS8L2 | |
| EPS8L2 Fusion Protein Ag14296 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction