missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS8L2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06508MFV450
This item is not returnable.
View return policy
Description
EPS8L2 Polyclonal antibody specifically detects EPS8L2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| EPS8L2 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| epidermal growth factor receptor kinase substrate 8-like protein 2, Epidermal growth factor receptor pathway substrate 8-related protein 2, EPS8-like 2, EPS8-like protein 2, EPS8R2, EPS8-related protein 2, FLJ16738, FLJ21935, FLJ22171, MGC126530, MGC3088 | |
| Produced in rabbits immunized with E. coli-derived Human EPS8L2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 64787 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction