missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS8L2 Antibody (6C2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064787-M01
This item is not returnable.
View return policy
Description
EPS8L2 Monoclonal antibody specifically detects EPS8L2 in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| EPS8L2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| epidermal growth factor receptor kinase substrate 8-like protein 2, Epidermal growth factor receptor pathway substrate 8-related protein 2, EPS8-like 2, EPS8-like protein 2, EPS8R2, EPS8-related protein 2, FLJ16738, FLJ21935, FLJ22171, MGC126530, MGC3088 | |
| EPS8L2 (NP_073609, 615 a.a. ∽ 714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGED | |
| 0.1 mg | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 6C2 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_073609 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 64787 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction