Learn More
Invitrogen™ EPS15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597253
Description
Immunogen sequence: RQSTDPFATS STDPFSAANN SSITSVETLK HNDPFAPGGT VVAASDSATD PFASVFGNES FGGGFADFST LSKVNNEDPF RSATSSSVSN VVITKNVFEE TSVKSEDEPP ALPPKIGTPT RPCPLPPGKR SINKLDSPDP FKLNDPFQPF PGNDSPKEKD PEIFCDPFTS ATTTTNKEAD P; Positive Samples: HeLa, Jurkat, 293T, NIH/3T3, Mouse liver, Mouse spleen, Mouse lung; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Early endosome membrane, Membrane, Peripheral membrane protein, clathrin-coated pit.
EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.
Specifications
| EPS15 | |
| Polyclonal | |
| Unconjugated | |
| EPS15 | |
| 2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; epidermal growth factor receptor substrate 15; EPS15; MLLT5; protein AF-1p; Protein Eps15; RGD1307641 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13858, 2060, 313474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42566, P42567 | |
| EPS15 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.