missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS15 Antibody (1261C) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8480JF635
This item is not returnable.
View return policy
Description
EPS15 Monoclonal antibody specifically detects EPS15 in Human samples. It is validated for Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifications
| EPS15 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
| E.coli-derived recombinant human Eps15, Ala448-Thr579, Accession # P42566 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1261C | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 2060 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction