missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS15 Antibody (1261C) [FITC], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8480F
This item is not returnable.
View return policy
Description
EPS15 Monoclonal antibody specifically detects EPS15 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| EPS15 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1261C | |
| Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
| E.coli-derived recombinant human Eps15, Ala448-Thr579, Accession # P42566 | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 2060 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction