missing translation for 'onlineSavingsMsg'
Learn More

Eprotirome, MedChemExpress

Product Code. 16411059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411059 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411059 Supplier Medchem Express Supplier No. HY10473
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eprotirome (KB2115) is a liver-selective thyroid hormone receptor (TR) agonist. KB2115 has modestly higher affinity for TRβ than for TRα. Eprotirome reduces low-density lipoprotein (LDL) cholesterol concentrations. Eprotirome can be used for dyslipidemias and obesity research.

Eprotirome (KB2115) is a liver-selective thyroid hormone receptor (TR) agonist. KB2115 has modestly higher affinity for TRβ than for TRα. Eprotirome reduces low-density lipoprotein (LDL) cholesterol concentrations. Eprotirome can be used for dyslipidemias and obesity research.

Specifications

Chemical Name or Material Eprotirome
CAS 355129-15-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H17Br2NO5
Quantity 1 mg
Synonym KB2115
Solubility Information DMSO : 125 mg/mL (256.60 mM; Need ultrasonic)
SMILES O=C(O)CC(NC1=CC(Br)=C(OC2=CC=C(O)C(C(C)C)=C2)C(Br)=C1)=O
Molecular Weight (g/mol) 487.14
Formula Weight 487.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.