missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eprosartan Mesylate, TRC
Specifications
Specifications
| Chemical Name or Material | Eprosartan Mesilate |
| CAS | 144143-96-4 |
| Molecular Formula | C23 H24 N2 O4 S . C H4 O3 S |
| Quantity | 10 mg |
| Synonym | 2-Thiophenepropanoic acid, α-[[2-butyl-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-, (E)-, monomethanesulfonate, 2-Thiophenepropanoic acid, α-[[2-butyl-1-[(4-carboxyphenyl)methyl]-1H-imidazol-5-yl]methylene]-, (αE)-, monomethanesulfonate, Eprosartan Mesylate, Eprosartan methanesulfonate, SKF 108566J |
| InChI Formula | InChI=1S/C23H24N2O4S.CH4O3S/c1-2-3-6-21-24-14-19(12-18(23(28)29)13-20-5-4-11-30-20)25(21)15-16-7-9-17(10-8-16)22(26)27;1-5(2,3)4/h4-5,7-12,14H,2-3,6,13,15H2,1H3,(H,26,27)(H,28,29);1H3,(H,2,3,4)/b18-12+; |
| SMILES | CCCCc1ncc(\C=C(/Cc2cccs2)\C(=O)O)n1Cc3ccc(cc3)C(=O)O.CS(=O)(=O)O |
| IUPAC Name | 4-[[2-butyl-5-[(E)-2-carboxy-3-thiophen-2-ylprop-1-enyl]imidazol-1-yl]methyl]benzoic acid;methanesulfonic acid |
| Molecular Weight (g/mol) | 520.62 |
| Formula Weight | 520.1338 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?