missing translation for 'onlineSavingsMsg'
Learn More

Eprosartan mesylate, MedChemExpress

Product Code. 16456858
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456858 10 mg 10mg
16446956 100 mg 100mg
16432927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456858 Supplier Medchem Express Supplier No. HY15834A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eprosartan mesylate (SKF-108566J) is a selective, competitive, nonpeptid and orally active angiotensin II receptor antagonist, used as an antihypertensive. Eprosartan mesylate binds angiotensin II receptor with IC50s of 9.2 nM and 3.9 nM in rat and human adrenal cortical membranes, respectively.

Eprosartan mesylate (SKF-108566J) is a selective, competitive, nonpeptid and orally active angiotensin II receptor antagonist, used as an antihypertensive. Eprosartan mesylate binds angiotensin II receptor with IC50s of 9.2 nM and 3.9 nM in rat and human adrenal cortical membranes, respectively.

Specifications

Chemical Name or Material Eprosartan mesylate
CAS 144143-96-4
Color White
Purity Grade Notes Research
Molecular Formula C24H28N2O7S2
Quantity 10 mg
Synonym SKF-108566J
Solubility Information DMSO : ≥ 48 mg/mL (92.20 mM)
SMILES OC(/C(CC1=CC=CS1)=C/C2=CN=C(CCCC)N2CC3=CC=C(C(O)=O)C=C3)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 520.62
Formula Weight 520.62
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.