missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eprosartan Acyl-Beta-D-glucuronide (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | Eprosartan Acyl-beta-D-glucuronide |
| Analyte or Component Names | Eprosartan Acyl-beta-D-glucuronide |
| Shipping Condition | Dry ice |
| Molecular Formula | C29 H32 N2 O10 S |
| InChI Formula | InChI=1S/C29H32N2O10S/c1-2-3-6-21-30-14-19(12-18(26(35)36)13-20-5-4-11-42-20)31(21)15-16-7-9-17(10-8-16)28(39)41-29-24(34)22(32)23(33)25(40-29)27(37)38/h4-5,7-12,14,22-25,29,32-34H,2-3,6,13,15H2,1H3,(H,35,36)(H,37,38)/b18-12+/t22-,23-,24+,25-,29-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCc1ncc(\C=C(/Cc2cccs2)\C(=O)O)n1Cc3ccc(cc3)C(=O)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[4-[[2-butyl-5-[(E)-2-carboxy-3-thiophen-2-ylprop-1-enyl]imidazol-1-yl]methyl]benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 600.637 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?