missing translation for 'onlineSavingsMsg'
Learn More

Eprosartan Acyl-Beta-D-glucuronide (>80%), TRC

Product Code. 30371214
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371214 0.5 mg 0.5mg
30371213 10 mg 10mg
30385805 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371214 Supplier Toronto Research Chemicals Supplier No. TRCE5901100.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eprosartan Acyl-beta-D-glucuronide
Analyte or Component Names Eprosartan Acyl-beta-D-glucuronide
Shipping Condition Dry ice
Molecular Formula C29 H32 N2 O10 S
InChI Formula InChI=1S/C29H32N2O10S/c1-2-3-6-21-30-14-19(12-18(26(35)36)13-20-5-4-11-42-20)31(21)15-16-7-9-17(10-8-16)28(39)41-29-24(34)22(32)23(33)25(40-29)27(37)38/h4-5,7-12,14,22-25,29,32-34H,2-3,6,13,15H2,1H3,(H,35,36)(H,37,38)/b18-12+/t22-,23-,24+,25-,29-/m0/s1
Solution Type Neat
SMILES CCCCc1ncc(\C=C(/Cc2cccs2)\C(=O)O)n1Cc3ccc(cc3)C(=O)O[C@@H]4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6S)-6-[4-[[2-butyl-5-[(E)-2-carboxy-3-thiophen-2-ylprop-1-enyl]imidazol-1-yl]methyl]benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 600.637
Quantity 0.5 mg
Formula Weight 600.178
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.