missing translation for 'onlineSavingsMsg'
Learn More

Eprazinone Dihydrochloride, TRC

Product Code. 30381739
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381739 50 mg
30381892 1 g
30371830 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381739 Supplier Toronto Research Chemicals Supplier No. TRCE59002050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10402-53-6
Molecular Formula C24H34Cl2N2O2
Molecular Weight (g/mol) 453.44
Synonym 3-(4-(2-Ethoxy-2-phenylethyl)piperazin-1-yl)-2-methyl-1-phenylpropan-1-one Dihydrochloride, 3-[4-(2-Ethoxy-2-phenylethyl)-1-piperazinyl]-2-methyl-1-phenyl-1-propanone Dihydrochloride, 3-[4-(β-Ethoxyphenethyl)-1-piperazinyl]-2-methylpropiophenone, Dihydrochloride, 3-[4-(2-Ethoxy-2-phenylethyl)-1-piperazinyl]-2-methyl-1-phenyl-1-propanone Dihydrochloride, 3-[4-(β-Ethoxyphenethyl)-1-piperazinyl]-2-methylpropiophenone Dihydrochloride, 746CE, Eftapan, Eprazinone Hydrochloride, Mucitux, NSC 317935, Resplen
SMILES Cl.Cl.CCOC(CN1CCN(CC(C)C(=O)c2ccccc2)CC1)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H32N2O2.2ClH/c1-3-28-23(21-10-6-4-7-11-21)19-26-16-14-25(15-17-26)18-20(2)24(27)22-12-8-5-9-13-22;;/h4-13,20,23H,3,14-19H2,1-2H3;2*1H
Chemical Name or Material Eprazinone Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.