missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EPPTB
GREENER_CHOICE

Product Code. 18770469
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770469 10 mg 10mg
18706168 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18770469 Supplier Tocris Bioscience™ Supplier No. 4518/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trace amine 1 (TA1) receptor antagonist/inverse agonist

EPPTB is a trace amine 1 (TA1) receptor antagonist/inverse agonist; exhibits a higher potency at the mouse TA1 receptor than the rat and human TA1 receptors (IC50 values are 27.5, 4539 and 7487 nM, respectively). Blocks the TA1 receptor-mediated activation of an inwardly rectifying K+ current, which increases the firing frequency of dopamine (DA) neurons in the ventral tegmental area. Increases DA potency at the D2 receptor. Also displays inverse agonism, reducing basal cAMP levels in vitro (IC50 = 19 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Ethoxyphenyl)-4-(1-pyrrolidinyl)-3-(trifluoromethyl)benzamide
CAS 1110781-88-8
Quantity 10 mg
Target Trace Amine 1 (TA1) Receptor Antagonists
Molecular Formula C20H21F3N2O2
Formula Weight 378.39
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.