missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EPPTB

Description
EPPTB is a trace amine 1 (TA1) receptor antagonist/inverse agonist; exhibits a higher potency at the mouse TA1 receptor than the rat and human TA1 receptors (IC50 values are 27.5, 4539 and 7487 nM, respectively). Blocks the TA1 receptor-mediated activation of an inwardly rectifying K+ current, which increases the firing frequency of dopamine (DA) neurons in the ventral tegmental area. Increases DA potency at the D2 receptor. Also displays inverse agonism, reducing basal cAMP levels in vitro (IC50 = 19 nM).
Specifications
Specifications
| Chemical Name or Material | N-(3-Ethoxyphenyl)-4-(1-pyrrolidinyl)-3-(trifluoromethyl)benzamide |
| CAS | 1110781-88-8 |
| Quantity | 50 mg |
| Target | Trace Amine 1 (TA1) Receptor Antagonists |
| Molecular Formula | C20H21F3N2O2 |
| Formula Weight | 378.39 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?