missing translation for 'onlineSavingsMsg'
Learn More

EPPS, JustPURE™, white powder, Fisher BioReagents

Product Code. 10222510
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10222510 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10222510 Supplier Fisher BioReagents Supplier No. BP2933100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Absorbance 0.1 max. at 280nm
CAS 16052-06-5
Melting Point 237°C
Color White
pH 5.0 to 6.0
Physical Form Powder or Solid
Quantity 100 g
Assay Percent Range ≥99.5 %
Molecular Formula C9H20N2O4S
Packaging Poly Bottle
Synonym 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid, HEPPS
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular Weight (g/mol) 252.329
PubChem CID 85255
ChEBI CHEBI:42298
Formula Weight 252.33
Percent Purity ≥99.5%
Grade JustPURE
Chemical Name or Material EPPS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.