missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Faster™ Epoxy Powder-coated Modular Stands for SafeFAST Safety Cabinets
Click to view available options
For Use With (Equipment):
SafeFAST Top 209
SafeFAST Top 212
SafeFAST Top 215
SafeFAST Top 218
Includes:
Stand
Stand with Castors
Description
- Stand with 800 mm height
- Made of epoxy-coated steel
- If included, lockable castors at the front
Specifications
Specifications
| For Use With (Equipment) | SafeFAST Top 215 |
| Includes | Stand |
| Color | White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction