missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Epoxomicin

Product Code. 16416346
Encompass_Preferred
Change view
Click to view available options
:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16416346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16416346 Supplier MedChemExpress Supplier No. HY13821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epoxomicin (BU-4061T) is an epoxyketone-containing natural product and a potent, selective and irreversible proteasome inhibitor. Epoxomicin covalently binds to the LMP7, X, MECL1, and Z catalytic subunits of the proteasome and potently inhibits primarily the chymotrypsin-like activity. Epoxomicin can cross the blood-brain barrier. Epoxomicin has strongly antitumor and anti-inflammatory activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 554.72 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 134381-21-8
Product Type Natural Products
Purity 95.0%
Solubility DMSO : 100 mg/mL (180.27 mM∣Need ultrasonic)
Synonym BU-4061T
Target Proteasome∣Apoptosis
Quantity 100 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.