missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Epoxomicin

Product Code. 16400556
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16400556 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16400556 Supplier MedChemExpress Supplier No. HY13821
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Epoxomicin (BU-4061T) is an epoxyketone-containing natural product and a potent, selective and irreversible proteasome inhibitor. Epoxomicin covalently binds to the LMP7, X, MECL1, and Z catalytic subunits of the proteasome and potently inhibits primarily the chymotrypsin-like activity. Epoxomicin can cross the blood-brain barrier. Epoxomicin has strongly antitumor and anti-inflammatory activity.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 554.72 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 134381-21-8
Product Type Natural Products
Purity 95.0%
Solubility DMSO : 100 mg/mL (180.27 mM∣Need ultrasonic)
Synonym BU-4061T
Target Proteasome∣Apoptosis
Quantity 20 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.