missing translation for 'onlineSavingsMsg'
Learn More

Epoxide Biperiden, TRC

Product Code. 30387117
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387117 25 mg 25mg
30372879 50 mg 50mg
30370061 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387117 Supplier Toronto Research Chemicals Supplier No. TRCE43918525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Epoxide Biperiden
Analyte or Component Names Epoxide Biperiden
Shipping Condition Room Temperature
Molecular Formula C21 H29 N O2
Synonym Biperiden Epoxide, 1-Phenyl-3-(piperidin-1-yl)-1-((1S,4R,5R,6R)-3-oxatricyclo[3.2.1.02,4]octan-6-yl)propan-1-ol
InChI Formula InChI=1S/C21H29NO2/c23-21(16-7-3-1-4-8-16,9-12-22-10-5-2-6-11-22)18-14-15-13-17(18)20-19(15)24-20/h1,3-4,7-8,15,17-20,23H,2,5-6,9-14H2/t15-,17+,18+,19?,20+,21?/m0/s1
Solution Type Neat
SMILES OC(CCN1CCCCC1)([C@@H]2C[C@@H]3C[C@H]2[C@H]4OC34)c5ccccc5
IUPAC Name 1-[(1S,4R,5R,6R)-3-oxatricyclo[3.2.1.02,4]octan-6-yl]-1-phenyl-3-piperidin-1-ylpropan-1-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 327.461
Quantity 25 mg
Formula Weight 327.22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.