missing translation for 'onlineSavingsMsg'
Learn More

Epothilone D (synthetic), TRC

Product Code. 30138412
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138412 25 mg 25mg
30133034 1 mg 1mg
30006339 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138412 Supplier Toronto Research Chemicals Supplier No. TRCE58900025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189453-10-9
Molecular Formula C26 H39 N O5 S
Molecular Weight (g/mol) 477.66
IUPAC Name (4S,7R,8S,9S,13Z,16S)-4,8-dihydroxy-5,5,7,9-tetramethyl-16-[(E)-1-(2-methyl-1,3-thiazol-4-yl)prop-1-en-2-yl]-1-oxacyclohexadec-13-ene-2,6-dione
SMILES C[C@H]1CCC\C=C/C[C@H](OC(=O)C[C@H](O)C(C)(C)C(=O)[C@H](C)[C@H]1O)\C(=C\c2csc(C)n2)\C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H39NO5S/c1-16-11-9-7-8-10-12-21(17(2)13-20-15-33-19(4)27-20)32-23(29)14-22(28)26(5,6)25(31)18(3)24(16)30/h8,10,13,15-16,18,21-22,24,28,30H,7,9,11-12,14H2,1-6H3/b10-8-,17-13+/t16-,18+,21-,22-,24-/m0/s1
Formula Weight 477.2549
Chemical Name or Material Epothilone D (synthetic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.