missing translation for 'onlineSavingsMsg'
Learn More

Epothilone A, TRC

Product Code. 30387653
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387653 1 mg
30380970 50 mg
30378572 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387653 Supplier Toronto Research Chemicals Supplier No. TRCE5889801MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152044-53-6
Molecular Formula C26H39NO6S
Molecular Weight (g/mol) 493.66
Synonym (1S,3S,7S,10R,11S,12S,16R)-7,11-Dihydroxy-8,8,10,12-tetramethyl-3-[(1E)-1-methyl-2-(2-methyl-4-thiazolyl)ethenyl]-4,17-dioxabicyclo[14.1.0]heptadecane-5,9-dione, (-)-Epothilone A, Epothilone A
SMILES C[C@@H]1CCC[C@@H]2O[C@@H]2C[C@@H](OC(=O)C[C@@H](O)C(C)(C)C(=O)[C@@H](C)[C@@H]1O)\C(=C\c3csc(C)n3)\C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H39NO6S/c1-14-8-7-9-19-21(32-19)11-20(15(2)10-18-13-34-17(4)27-18)33-23(29)12-22(28)26(5,6)25(31)16(3)24(14)30/h10,13-14,16,19-22,24,28,30H,7-9,11-12H2,1-6H3/b15-10+/t14-,16+,19+,20-,21-,22-,24-/m0/s1
Chemical Name or Material Epothilone A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.