missing translation for 'onlineSavingsMsg'
Learn More

Epothilone A, MedChemExpress

Product Code. 16491349
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491349 1 mg 1mg
16458407 5 mg 5mg
16435718 10 mg 10mg
16433366 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491349 Supplier Medchem Express Supplier No. HY13503
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Epothilone A is a competitive inhibitor of the binding of [3H] paclitaxel to tubulin polymers, with a Ki of 0.6-1.4 μM.

Epothilone A is a competitive inhibitor of the binding of [3H] paclitaxel to tubulin polymers, with a Ki of 0.6-1.4 μM.

Specifications

Chemical Name or Material Epothilone A
Physical Form Solid
CAS 152044-53-6
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C26H39NO6S
For Use With (Application) Cancer-programmed cell death
Synonym Epo A
Solubility Information DMSO : 125 mg/mL (253.21 mM; Need ultrasonic)
SMILES O=C(C[C@H](O)C1(C)C)O[C@H](/C(C)=C/C2=CSC(C)=N2)C[C@]3([H])O[C@]3([H])CCC[C@H](C)[C@H](O)[C@@H](C)C1=O
Molecular Weight (g/mol) 493.66
Formula Weight 493.66
Percent Purity 98.61%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.