missing translation for 'onlineSavingsMsg'
Learn More

Epostane, TRC

Product Code. 30382167
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382167 5 mg
30382795 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382167 Supplier Toronto Research Chemicals Supplier No. TRCE5888505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80471-63-2
Molecular Formula C22 H31 N O3
Molecular Weight (g/mol) 357.49
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@@]45O[C@]4(C)C(=C(C[C@]5(C)[C@H]3CC[C@]12C)C#N)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H31NO3/c1-18-8-6-16-14(15(18)7-9-20(18,3)25)5-10-22-19(16,2)11-13(12-23)17(24)21(22,4)26-22/h14-16,24-25H,5-11H2,1-4H3/t14-,15-,16-,18-,19+,20-,21+,22-/m0/s1
Formula Weight 357.2304
Chemical Name or Material Epostane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.